Gene Transcripts

Gene Symbol
Stxbp4
Entrez Gene ID
20913
Gene Name
syntaxin binding protein 4
Gene Aliases
6030470M02Rik, Synip
Chromosome Location
Chr. 11: 90476492 - 90638123 on Build GRCm38
UniGene ID
Mm.207203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011505.2 NP_035635.1 17 1772
GenBank mRNA AF152924.1 - 17 1762
AK039029.1 - 5 446
AK138499.1 - 5 445

View Full Product Details