Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001567.3 | NP_001558.3 | 21 | 2489 | |
XM_005273978.3 | XP_005274035.1 | 22 | 2866 | ||
XM_005273979.4 | XP_005274036.1 | 22 | 2713 | ||
XM_011544999.2 | XP_011543301.1 | 22 | 2647 | ||
XM_011545000.2 | XP_011543302.1 | 22 | 2713 | ||
XM_017017673.1 | XP_016873162.1 | 23 | 2804 | ||
GenBank mRNA | BC140853.1 | - | 21 | 2489 | |
L36818.1 | - | 20 | 2466 | ||
Y14385.1 | - | 21 | 2553 |