Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011528.4 | NP_035658.1 | 3, 4 | 400 | |
GenBank mRNA | AK003092.1 | - | 3, 4 | 362 | |
AK142030.1 | - | 3, 4 | 358 | ||
BC003970.1 | - | 3, 4 | 333 | ||
BC004754.1 | - | 3, 4 | 355 | 3584511 | |
BC094277.1 | - | 3, 4 | 349 | 5008399 | |
U67611.1 | - | 3, 4 | 343 |