Gene Transcripts

Gene Symbol
Tlx1
Entrez Gene ID
21908
Gene Name
T cell leukemia, homeobox 1
Gene Aliases
Hox-11, Hox11
Chromosome Location
Chr. 19: 45150079 - 45156943 on Build GRCm38
UniGene ID
Mm.391203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021901.3 NP_068701.1 1 268
GenBank mRNA BC018246.1 - 1 268

View Full Product Details