Gene Transcripts

Gene Symbol
Cd70
Entrez Gene ID
21948
Gene Name
CD70 antigen
Gene Aliases
CD27LG, Cd27l, Tnfsf7
Chromosome Location
Chr. 17: 57145997 - 57149777 on Build GRCm38
UniGene ID
Mm.42228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011617.2 NP_035747.1 3 494
GenBank mRNA BC125407.1 - 3 453
BC125409.1 - 3 453 40129924
U78091.1 - 3 376
Y13636.1 - 3 494

View Full Product Details