Gene Transcripts
- Gene Symbol
- Tom1
- Entrez Gene ID
- 21968
- Gene Name
- target of myb1 homolog (chicken)
- Gene Aliases
- Chromosome Location
- Chr. 8: 75033686 - 75070121 on Build GRCm38
- UniGene ID
- Mm.1967
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001136259.1 |
NP_001129731.1
|
5, 6 |
598 |
|
NM_011622.3 |
NP_035752.1
|
5, 6 |
598 |
|
GenBank mRNA |
AJ006972.1 |
- |
5, 6 |
575 |
|
AK028398.1 |
- |
5, 6 |
539 |
|
AK051245.1 |
- |
5, 6 |
580 |
|
AK150143.1 |
- |
5, 6 |
548 |
|
AK160403.1 |
- |
5, 6 |
544 |
|
BC021633.1 |
- |
5, 6 |
561 |
4208226 |
BC093520.1 |
- |
5, 6 |
526 |
2866013 |
BC157905.1 |
- |
5, 6 |
559 |
9088106 |
BC158042.1 |
- |
5, 6 |
559 |
9007343 |
View Full Product Details