Gene Transcripts
- Gene Symbol
- Tpi1
- Entrez Gene ID
- 21991
- Gene Name
- triosephosphate isomerase 1
- Gene Aliases
- AI255506, Tpi, Tpi-1
- Chromosome Location
- Chr. 6: 124810592 - 124814296 on Build GRCm38
- UniGene ID
- Mm.4222
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009415.2 |
NP_033441.2
|
4, 5 |
625 |
|
GenBank mRNA |
AK002270.1 |
- |
4, 5 |
487 |
|
AK010808.1 |
- |
4, 5 |
495 |
|
AK146013.1 |
- |
4, 5 |
484 |
|
AK149768.1 |
- |
4, 5 |
489 |
|
AK150735.1 |
- |
4, 5 |
486 |
|
AK159741.1 |
- |
4, 5 |
489 |
|
AK167437.1 |
- |
4, 5 |
490 |
|
AK168060.1 |
- |
4, 5 |
487 |
|
AK168446.1 |
- |
4, 5 |
489 |
|
AK168756.1 |
- |
4, 5 |
488 |
|
BC004024.1 |
- |
4, 5 |
473 |
|
BC046761.1 |
- |
4, 5 |
476 |
5693696 |
BU945752.1 |
- |
4, 5 |
631 |
|
X53333.1 |
- |
4, 5 |
487 |
|
View Full Product Details