Gene Transcripts

Gene Symbol
Tpmt
Entrez Gene ID
22017
Gene Name
thiopurine methyltransferase
Gene Aliases
AW106912
Chromosome Location
Chr. 13: 47021987 - 47043338 on Build GRCm38
UniGene ID
Mm.10169
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF037043.1 - 2, 3 206
AF037045.1 - 1, 2 157
AF046887.1 - 2, 3 283
BC021598.1 - 1, 2 184 5098857
CF106558.1 - 2, 3 219

View Full Product Details