Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011639.3 | NP_035769.1 | 8 | 1464 | |
GenBank mRNA | AF097511.1 | - | 8 | 1357 | |
AF116823.1 | - | 8 | 1459 | ||
AK010435.1 | - | 8 | 1465 | ||
AK146823.1 | - | 8 | 1414 | ||
AK159387.1 | - | 8 | 1433 | ||
AK166024.1 | - | 8 | 1628 | ||
BC138685.1 | - | 8 | 1396 | 8861706 | |
BC138686.1 | - | 8 | 1396 | 8861707 |