Gene Transcripts

Gene Symbol
Trpc4
Entrez Gene ID
22066
Gene Name
transient receptor potential cation channel, subfamily C, member 4
Gene Aliases
CCE1, STRPC4, Trp4, Trrp4
Chromosome Location
Chr. 3: 54156057 - 54318471 on Build GRCm38
UniGene ID
Mm.10100
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253682.1 NP_001240611.1 5 1590
NM_016984.3 NP_058680.1 5 1590
GenBank mRNA AF011543.1 - 5 1407
AF019663.1 - 5 1407
AF190646.1 - 5 1481
AK044265.1 - 5 1446
AK080670.1 - 5 1591
BC057335.1 - 5 1426
BC060160.1 - 5 1426
BC120729.1 - 5 1505 40129652
BC131915.1 - 5 1505 40130192
U50921.1 - 5 1543
U50922.1 - 5 1543

View Full Product Details