Gene Transcripts

Gene Symbol
Ugt8a
Entrez Gene ID
22239
Gene Name
UDP galactosyltransferase 8A
Gene Aliases
AI850488, AW455908, Cgt, Ugt8, mCerGT
Chromosome Location
Chr. 3: 125865322 - 125938550 on Build GRCm38
UniGene ID
Mm.306021
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011674.4 NP_035804.2 2 328
GenBank mRNA AK078371.1 - 2 330
AK128994.1 - 2 324
AK137364.1 - 2 329
AK144005.1 - 2 329
AK163650.1 - 2 290
BC016885.1 - 323 4223057
X92122.1 - 1 653

View Full Product Details