Gene Transcripts
- Gene Symbol
- Usp4
- Entrez Gene ID
- 22258
- Gene Name
- ubiquitin specific peptidase 4 (proto-oncogene)
- Gene Aliases
- F730026I20Rik, Unp, mKIAA4155
- Chromosome Location
- Chr. 9: 108347820 - 108392529 on Build GRCm38
- UniGene ID
- Mm.3974
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011678.2 |
NP_035808.2
|
10, 11 |
1375 |
|
GenBank mRNA |
AK143582.1 |
- |
10, 11 |
1310 |
|
AK149964.1 |
- |
10, 11 |
1333 |
|
AK164245.1 |
- |
1, 2 |
150 |
|
AK169372.1 |
- |
10, 11 |
1354 |
|
AK169933.1 |
- |
10, 11 |
1345 |
|
AK171271.1 |
- |
10, 11 |
1345 |
|
AK171633.1 |
- |
10, 11 |
1328 |
|
AK220512.1 |
- |
10, 11 |
1332 |
|
BC011341.1 |
- |
9, 10 |
1152 |
3661165 |
BC066180.1 |
- |
10, 11 |
1285 |
|
BC066865.1 |
- |
10, 11 |
1285 |
|
L00681.1 |
- |
10, 11 |
1338 |
|
View Full Product Details