Gene Transcripts

Gene Symbol
Zfp35
Entrez Gene ID
22694
Gene Name
zinc finger protein 35
Gene Aliases
Zfp-35, Znf271, mszf13, mszf20-1, mszf28, mszf68
Chromosome Location
Chr. 18: 23989634 - 24005371 on Build GRCm38
UniGene ID
Mm.290596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011755.2 NP_035885.2 3 1373
GenBank mRNA AK132703.1 - 3 1376
AK159080.1 - 3 1327
BC138830.1 - 3 1292 8861852
BC138832.1 - 2 1290 8861854
X17617.1 - 3 1324

View Full Product Details