Gene Transcripts

Gene Symbol
Zfp51
Entrez Gene ID
22709
Gene Name
zinc finger protein 51
Gene Aliases
Zfp-51, zfec12
Chromosome Location
Chr. 17: 21450352 - 21465591 on Build GRCm38
UniGene ID
Mm.293030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK133677.1 - 5 1373
AK154157.1 - 5 1379
BC011183.1 - 4 1341 4036027
X74855.1 - 5 1404

View Full Product Details