Gene Transcripts

Gene Symbol
Zfy2
Entrez Gene ID
22768
Gene Name
zinc finger protein 2, Y linked
Gene Aliases
Zfy-2
Chromosome Location
Chr. Y: 2106015 - 2170409 on Build GRCm38
UniGene ID
Mm.411381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009571.2 NP_033597.2 3 408
GenBank mRNA AK030048.1 - 3 279
AK133222.1 - 3 409

View Full Product Details