Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145947.1 | NP_001139419.1 | 5 | 1308 | |
NM_011792.5 | NP_035922.4 | 5 | 1308 | ||
GenBank mRNA | AF190726.2 | - | 5 | 1236 | |
AF200346.1 | - | 5 | 809 | ||
AK014390.2 | - | 1 | 107 | ||
AK014464.1 | - | 5 | 1238 | ||
AK033112.1 | - | 5 | 1258 | ||
AK041285.1 | - | 5 | 1258 | ||
AK046175.1 | - | 5 | 1255 | ||
AK049626.1 | - | 5 | 1241 | ||
AK079814.1 | - | 4 | 526 | ||
AK080498.1 | - | 5 | 1260 | ||
AK082230.1 | - | 5 | 1257 | ||
AK082317.1 | - | 5 | 1259 | ||
AK163583.1 | - | 5 | 1259 | ||
AK173112.1 | - | 3 | 426 | ||
AK190595.1 | - | 1 | 14 | ||
AK197470.1 | - | 177 | |||
BC048189.1 | - | 5 | 1234 | 6831622 |