Gene Transcripts

Gene Symbol
Banf1
Entrez Gene ID
23825
Gene Name
barrier to autointegration factor 1
Gene Aliases
Baf, Bcrp1, L2bp1/Baf
Chromosome Location
Chr. 19: 5364633 - 5366876 on Build GRCm38
UniGene ID
Mm.358649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038231.2 NP_001033320.1 3 268
NM_001286608.1 NP_001273537.1 3 610
NM_011793.3 NP_035923.1 3 476
GenBank mRNA AB025349.1 - 3 208
AF037080.1 - 2 151
AK003025.1 - 3 252
AK004212.1 - 3 235
AK010257.1 - 3 254
AK011079.1 - 3 257
AK011100.1 - 3 255
AK012588.1 - 3 238
AK088896.1 - 3 232
AK168039.1 - 3 257
BC003709.1 - 3 245 3599828
CK780850.1 - 3 240

View Full Product Details