Gene Transcripts

Gene Symbol
Insrr
Entrez Gene ID
23920
Gene Name
insulin receptor-related receptor
Gene Aliases
Irr
Chromosome Location
Chr. 3: 87796908 - 87816101 on Build GRCm38
UniGene ID
Mm.42041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011832.2 NP_035962.2 14 3191
GenBank mRNA AB007135.1 - 14 3080
BC137855.1 - 14 3048 8860874

View Full Product Details