Gene Transcripts

Gene Symbol
Krtap15
Entrez Gene ID
26560
Gene Name
keratin associated protein 15
Gene Aliases
Krtap15-1, Pmg-2, Pmg2
Chromosome Location
Chr. 16: 88829009 - 88829844 on Build GRCm38
UniGene ID
Mm.86430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013713.1 NP_038741.1 1 403
GenBank mRNA AK132534.1 - 1 419
BC116219.2 - 1 402 40091298

View Full Product Details