Gene Transcripts

Gene Symbol
Abcb11
Entrez Gene ID
27413
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 11
Gene Aliases
ABC16, Bsep, Lith1, PFIC2, PGY4, SPGP
Chromosome Location
Chr. 2: 69238282 - 69342842 on Build GRCm38
UniGene ID
Mm.439855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021022.3 NP_066302.2 12 1363
GenBank mRNA AF133903.1 - 12 1334
AK144553.1 - 13 1438

View Full Product Details