Gene Transcripts

Gene Symbol
Amot
Entrez Gene ID
27494
Gene Name
angiomotin
Gene Aliases
CAG-2, D0Kist1, Sii6
Chromosome Location
Chr. X: 145446424 - 145505541 on Build GRCm38
UniGene ID
Mm.100068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153319.3 NP_695231.3 10 3247
GenBank mRNA AF461135.1 - 10 2648
AK129277.1 - 5 1487
U80888.1 - 1 819

View Full Product Details