Gene Transcripts

Gene Symbol
Nelfe
Entrez Gene ID
27632
Gene Name
negative elongation factor complex member E, Rdbp
Gene Aliases
D17H6S45, NELF-E, RD, Rdbp
Chromosome Location
Chr. 17: 34850391 - 34856372 on Build GRCm38
UniGene ID
Mm.279907
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045863.1 NP_001039328.1 4, 5 495
NM_001045864.1 NP_001039329.1 4, 5 581
NM_138580.2 NP_613046.2 4, 5 369
GenBank mRNA AK011663.1 - 4, 5 353
AK012389.1 - 4, 5 386
AK131947.1 - 4, 5 353
AK147011.1 - 4, 5 477
BC034867.1 - 4, 5 321 1495547
CN525174.1 - 4, 5 324
DV662511.1 - 1, 2 114

View Full Product Details