Gene Transcripts
- Gene Symbol
- Slco1a4
- Entrez Gene ID
- 28250
- Gene Name
- solute carrier organic anion transporter family, member 1a4
- Gene Aliases
- AI785519, Oatp1a4, Oatp2, Slc21a5
- Chromosome Location
- Chr. 6: 141805438 - 141856283 on Build GRCm38
- UniGene ID
- Mm.255586
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030687.1 |
NP_109612.1
|
11, 12 |
1368 |
|
GenBank mRNA |
AB031814.2 |
- |
11, 12 |
1368 |
|
View Full Product Details