Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004518.5 | NP_004509.2 | 3 | 701 | |
NM_172106.2 | NP_742104.1 | 3 | 701 | ||
NM_172107.3 | NP_742105.1 | 3 | 701 | ||
NM_172108.4 | NP_742106.1 | 3 | 701 | ||
NM_172109.2 | NP_742107.1 | 3 | 701 | ||
XM_011528810.1 | XP_011527112.1 | 3 | 613 | ||
XM_011528811.1 | XP_011527113.1 | 3 | 613 | ||
XM_017027841.1 | XP_016883330.1 | 3 | 613 | ||
XM_017027842.1 | XP_016883331.1 | 3 | 613 | ||
XM_017027843.1 | XP_016883332.1 | 3 | 506 | ||
XM_017027844.1 | XP_016883333.1 | 3 | 613 | ||
GenBank mRNA | AF033348.1 | - | 3 | 614 | |
AF074247.1 | - | 3 | 662 | ||
AF110020.1 | - | 3 | 533 | ||
AK293727.1 | - | 3 | 613 | ||
BC000699.2 | - | 3 | 530 | ||
BT007043.1 | - | 3 | 487 | ||
D82346.1 | - | 3 | 664 | ||
Y15065.1 | - | 3 | 529 |