Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013875.5 | NP_038903.3 | 2, 3 | 458 | |
GenBank mRNA | AF190639.1 | - | 2, 3 | 239 | |
AJ251859.1 | - | 2, 3 | 510 | ||
AK020137.1 | - | 2, 3 | 421 | ||
AK035385.1 | - | 2, 3 | 470 | ||
BC130267.1 | - | 2, 3 | 339 | 40143060 |