Gene Transcripts

Gene Symbol
Ebi3
Entrez Gene ID
50498
Gene Name
Epstein-Barr virus induced gene 3
Gene Aliases
EBI-3, IL-27
Chromosome Location
Chr. 17: 55952623 - 55957022 on Build GRCm38
UniGene ID
Mm.256798
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015766.2 NP_056581.1 5 866
GenBank mRNA AF013114.1 - 5 849
AK155023.1 - 5 829
AK155903.1 - 5 721
BC008209.1 - 5 814 3154957

View Full Product Details