Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015792.1 | NP_056607.1 | 17 | 2529 | |
GenBank mRNA | AF184275.1 | - | 15 | 2023 | |
AK003964.1 | - | 2 | 90 | ||
AK083118.1 | - | 17 | 2874 | ||
AK084645.1 | - | 17 | 2466 | ||
AK147829.1 | - | 17 | 2513 | ||
AK154357.1 | - | 17 | 2552 | ||
AK158937.1 | - | 17 | 2580 | ||
AK181430.1 | - | 3 | 160 | ||
AK211398.1 | - | 4 | 261 | ||
BC004036.1 | - | 14 | 1615 | ||
BC031393.1 | - | 17 | 2529 | 4948804 |