Gene Transcripts

Gene Symbol
Lsm4
Entrez Gene ID
50783
Gene Name
LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 8: 70673231 - 70678752 on Build GRCm38
UniGene ID
Mm.248188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015816.4 NP_056631.2 4 432
GenBank mRNA AJ249439.1 - 4 272
AK003847.1 - 4 268
AK010910.1 - 4 268
AK089028.1 - 4 285
AK143470.1 - 4 256
AK167784.1 - 4 296
AK185496.1 - 3 132
CA322716.1 - 4 432

View Full Product Details