Gene Transcripts

Gene Symbol
Neu3
Entrez Gene ID
50877
Gene Name
neuraminidase 3
Gene Aliases
Chromosome Location
Chr. 7: 99811439 - 99828525 on Build GRCm38
UniGene ID
Mm.103703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016720.2 NP_057929.1 3 1301
GenBank mRNA AB026842.1 - 3 1300
AK050428.1 - 3 1179
AK134405.1 - 3 1258
BC116892.1 - 3 1090 40126211
BC116924.1 - 3 1090 40126243

View Full Product Details