Gene Transcripts

Gene Symbol
Irx4
Entrez Gene ID
50916
Gene Name
Iroquois related homeobox 4 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 13: 73259590 - 73269622 on Build GRCm38
UniGene ID
Mm.103784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018885.2 NP_061373.1 6 1258
GenBank mRNA AF124732.1 - 6 963
AK132571.1 - 6 1258
BC126942.1 - 6 896 40089986

View Full Product Details