Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322168.1 | NP_001309097.1 | 3 | 314 | |
NM_013289.2 | NP_037421.2 | 3 | 314 | ||
GenBank mRNA | AF022049.1 | - | 3 | 251 | |
AF103010.1 | - | 3 | 251 | ||
AF103011.1 | - | 3 | 251 | ||
AF103012.1 | - | 3 | 251 | ||
AF103013.1 | - | 3 | 251 | ||
AF208688.1 | - | 1 | 188 | ||
AF208689.1 | - | 1 | 188 | ||
AF262968.1 | - | 1 | 170 | ||
AF262969.1 | - | 1 | 170 | ||
AF262970.1 | - | 1 | 170 | ||
AF262971.1 | - | 1 | 170 | ||
AF262972.1 | - | 1 | 170 | ||
AF262973.1 | - | 3 | 284 | ||
AF262974.1 | - | 3 | 284 | ||
AJ417556.1 | - | 1 | 200 | ||
AJ417557.1 | - | 1 | 200 | ||
AY366236.1 | - | 3 | 289 | ||
AY366249.1 | - | 3 | 251 | ||
AY366250.1 | - | 3 | 277 | ||
AY366251.1 | - | 3 | 251 | ||
AY366252.1 | - | 3 | 251 | ||
AY760050.1 | - | 3 | 251 | ||
AY789065.1 | - | 3 | 266 | ||
AY789066.1 | - | 3 | 251 | ||
AY789072.1 | - | 3 | 251 | ||
BC028206.1 | - | 3 | 278 | 5215674 | |
BC101540.1 | - | 3 | 270 | 8069046 | |
BC101542.1 | - | 3 | 270 | 8069048 | |
DQ178659.1 | - | 3 | 251 | ||
EF462885.1 | - | 1 | 181 | ||
EF472674.1 | - | 1 | 181 | ||
EF472675.1 | - | 1 | 181 | ||
EF472676.1 | - | 1 | 181 | ||
EF472677.1 | - | 1 | 181 | ||
EF472678.1 | - | 1 | 181 | ||
EF472679.1 | - | 1 | 181 | ||
L41269.1 | - | 3 | 270 | ||
U30273.1 | - | 3 | 253 | ||
U30274.1 | - | 3 | 314 | ||
U31416.1 | - | 3 | 276 | ||
U33328.1 | - | 3 | 276 | ||
X94262.1 | - | 3 | 252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083539.2 | NP_001077008.1 | 3 | 288 | |
GenBank mRNA | AF022044.1 | - | 3 | 264 | |
AJ417558.3 | - | 1 | 200 | ||
AK301140.1 | - | 3 | 288 | ||
BC098288.1 | - | 3 | 262 | 40026980 | |
BC105679.1 | - | 3 | 262 | 40004313 | |
DC390157.1 | - | 3 | 288 | ||
EF370406.1 | - | 2 | 212 | ||
EF582383.1 | - | 3 | 251 | ||
L76661.1 | - | 3 | 262 | ||
U73396.1 | - | 3 | 251 | ||
X97233.1 | - | 3 | 251 |