Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033525.3 | NP_001028697.2 | 2 | 616 | |
GenBank mRNA | AK036882.1 | - | 2 | 571 | |
AK132480.1 | - | 2 | 617 | ||
AK163847.1 | - | 2 | 465 | ||
AK171038.1 | - | 2 | 610 | ||
BC115791.1 | - | 2 | 322 | 40100611 | |
BC115915.1 | - | 2 | 316 | 40100605 | |
DQ185135.1 | - | 2 | 577 |