Gene Transcripts

Gene Symbol
Rragd
Entrez Gene ID
52187
Gene Name
Ras-related GTP binding D
Gene Aliases
5730543C08Rik, AI467523, C030003H22Rik, D4Ertd174e
Chromosome Location
Chr. 4: 32982998 - 33022180 on Build GRCm38
UniGene ID
Mm.300814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027491.2 NP_081767.2 2 691
GenBank mRNA AK017818.1 - 1 51
CK387674.1 - 2 270

View Full Product Details