Gene Transcripts

Gene Symbol
Ncald
Entrez Gene ID
52589
Gene Name
neurocalcin delta
Gene Aliases
AI848120, D030020D09Rik, D15Ertd412e
Chromosome Location
Chr. 15: 37366175 - 37792519 on Build GRCm38
UniGene ID
Mm.283370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170866.1 NP_001164337.1 5 544
NM_001170867.1 NP_001164338.1 3 478
NM_001170868.1 NP_001164339.1 2 426
NM_134094.4 NP_598855.2 5 734
GenBank mRNA AI893360.1 - 5 540
AK034031.1 - 3 457
AK048440.1 - 5 714
AK167313.1 - 2 396
BC011162.1 - 2 421 4191195
BC026979.1 - 2 380 5365560

View Full Product Details