Gene Transcripts

Gene Symbol
Plrg1
Entrez Gene ID
53317
Gene Name
pleiotropic regulator 1, PRL1 homolog (Arabidopsis)
Gene Aliases
AA958940, C80566, Tango4
Chromosome Location
Chr. 3: 83055538 - 83072291 on Build GRCm38
UniGene ID
Mm.286349
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016784.3 NP_058064.2 7 620
GenBank mRNA AF044334.1 - 6 512
AK075993.1 - 7 608
AK076007.1 - 7 598
AK077197.1 - 7 621
AK080068.1 - 7 627
BC006750.1 - 7 590 3709134
BC052895.1 - 7 578

View Full Product Details