Gene Transcripts
- Gene Symbol
- Timm23
- Entrez Gene ID
- 53600
- Gene Name
- translocase of inner mitochondrial membrane 23
- Gene Aliases
- Tim23
- Chromosome Location
- Chr. 14: 32180166 - 32201891 on Build GRCm38
- UniGene ID
- Mm.303703
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016897.3 |
NP_058593.2
|
2, 3 |
302 |
|
GenBank mRNA |
AB021122.1 |
- |
2, 3 |
282 |
|
AK013976.1 |
- |
2, 3 |
248 |
|
AK146516.1 |
- |
2, 3 |
268 |
|
BC037022.1 |
- |
2, 3 |
279 |
5366968 |
BC062963.1 |
- |
2, 3 |
238 |
5692352 |
BC069903.1 |
- |
2, 3 |
272 |
5290493 |
BC096669.1 |
- |
2, 3 |
271 |
5056242 |
BC132019.1 |
- |
2, 3 |
251 |
40130296 |
BC132319.1 |
- |
2, 3 |
251 |
40130596 |
BY767660.1 |
- |
2, 3 |
302 |
|
View Full Product Details