Gene Transcripts

Gene Symbol
Isg15
Entrez Gene ID
100038882
Gene Name
ISG15 ubiquitin-like modifier
Gene Aliases
G1p2, IGI15, IP17, Irfp, UCRP
Chromosome Location
Chr. 4: 156199424 - 156200818 on Build GRCm38
UniGene ID
Mm.451527
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015783.3 NP_056598.2 2 321
GenBank mRNA BC031424.1 - 2 288
BC083156.1 - 2 270 5338191
BC109346.2 - 2 271 40055493
CT010333.1 - 1 211
X56602.1 - 2 322

View Full Product Details