Gene Transcripts

Gene Symbol
Insm1
Entrez Gene ID
53626
Gene Name
insulinoma-associated 1
Gene Aliases
IA-1
Chromosome Location
Chr. 2: 146221997 - 146225020 on Build GRCm38
UniGene ID
Mm.379070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016889.3 NP_058585.2 1 2369
GenBank mRNA AF044669.1 - 1 2273
BC051240.1 - 1 2445 6469481
BC082809.1 - 1 2255 5720418

View Full Product Details