Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111314.1 | NP_001104784.1 | 5 | 846 | |
NM_019867.2 | NP_063920.2 | 3 | 483 | ||
GenBank mRNA | AJ238898.1 | - | 3 | 464 | |
AK138433.1 | - | 3 | 453 | ||
AY038025.1 | - | 3 | 458 | ||
BC039279.1 | - | 5 | 846 | 4973826 |