Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016867.1 | NP_058563.1 | 2 | 430 | |
GenBank mRNA | AF061262.1 | - | 2 | 430 | |
AK008307.1 | - | 2 | 247 | ||
AK008626.1 | - | 2 | 443 | ||
AK008771.1 | - | 2 | 762 | ||
AK168560.1 | - | 2 | 441 | ||
BC013440.1 | - | 2 | 484 | 4238716 |