Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164186.1 | NP_001157658.1 | 2, 3 | 925 | |
NM_019446.4 | NP_062319.1 | 1, 2 | 1208 | ||
GenBank mRNA | AB043980.1 | - | 1, 2 | 1223 | |
AF264026.1 | - | 3, 4 | 983 | ||
AJ237590.1 | - | 1, 2 | 483 | ||
AK036312.1 | - | 1, 2 | 660 | ||
BC055731.1 | - | 2, 3 | 926 | 6413504 |