Gene Transcripts

Gene Symbol
Nphs1
Entrez Gene ID
54631
Gene Name
nephrosis 1, nephrin
Gene Aliases
NephrinB, nephrin
Chromosome Location
Chr. 7: 30458206 - 30488609 on Build GRCm38
UniGene ID
Mm.437830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019459.2 NP_062332.2 22 3126
GenBank mRNA AB513652.1 - 22 2998
AF168466.1 - 22 2998
AF172256.1 - 22 3126
AF191090.1 - 22 3454
AK141081.1 - 22 3149

View Full Product Details