Gene Transcripts

Gene Symbol
Crtam
Entrez Gene ID
54698
Gene Name
cytotoxic and regulatory T cell molecule
Gene Aliases
Chromosome Location
Chr. 9: 40972798 - 41004628 on Build GRCm38
UniGene ID
Mm.42791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281954.1 NP_001268883.1 4 437
NM_019465.4 NP_062338.3 4 437
GenBank mRNA AK005719.1 - 2 160

View Full Product Details