Gene Transcripts

Gene Symbol
Mpc1
Entrez Gene ID
55951
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
0610006G08Rik, 3830411I18Rik, Brp44l
Chromosome Location
Chr. 17: 8283813 - 8297667 on Build GRCm38
UniGene ID
Mm.288510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018819.4 NP_061289.1 3, 4 261
GenBank mRNA AF181116.1 - 3, 4 217
AK002241.1 - 3, 4 262
AK002266.1 - 3, 4 240
AK004013.1 - 3, 4 217
AK005348.1 - 3, 4 253
AK013391.1 - 3, 4 262
AK014435.1 - 3, 4 491
AK135943.1 - 3, 4 232
AK170514.1 - 3, 4 278
BC024365.1 - 3, 4 223 4988147
BC103767.1 - 3, 4 264 30840199

View Full Product Details