Gene Transcripts

Gene Symbol
Ift20
Entrez Gene ID
55978
Gene Name
intraflagellar transport 20
Gene Aliases
0610009H04Rik, AU015496
Chromosome Location
Chr. 11: 78536436 - 78541473 on Build GRCm38
UniGene ID
Mm.358671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018854.4 NP_061342.1 4 524
GenBank mRNA AK002397.1 - 4 525
AK139942.1 - 4 561
AY082613.1 - 3 244
BC002040.1 - 4 515 3590790
U38981.1 - 357

View Full Product Details