Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291161.1 | NP_001278090.1 | 2 | 318 | |
NM_019818.5 | NP_062792.2 | 2 | 399 | ||
GenBank mRNA | AF223950.1 | - | 2 | 365 | |
AK011745.1 | - | 2 | 351 | ||
AK012130.2 | - | 2 | 353 | ||
AK017389.1 | - | 2 | 352 | ||
AK028132.1 | - | 2 | 351 | ||
AK153837.1 | - | 2 | 354 | ||
BC022610.1 | - | 2 | 335 | ||
BC055279.1 | - | 2 | 363 | 5688754 | |
BF470824.1 | - | 2 | 375 | ||
CA751902.1 | - | 2 | 291 | ||
Z49228.1 | - | 2 | 125 |