Gene Transcripts

Gene Symbol
Morf4l2
Entrez Gene ID
56397
Gene Name
mortality factor 4 like 2
Gene Aliases
2410017O14Rik, Mrgx, Sid393p, mKIAA0026
Chromosome Location
Chr. X: 136732942 - 136743686 on Build GRCm38
UniGene ID
Mm.27218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168225.1 NP_001161697.1 4 934
NM_001168226.1 NP_001161698.1 4 867
NM_001168227.1 NP_001161699.1 4 819
NM_001168228.1 NP_001161700.1 4 818
NM_001168229.1 NP_001161701.1 3 790
NM_001168230.1 NP_001161702.1 3 728
NM_019768.4 NP_062742.4 4 941
GenBank mRNA AB025049.1 - 2 629
AK075946.1 - 4 794
AK088849.1 - 1 648
AK089963.1 - 4 795
AK131944.1 - 4 794
AK145686.1 - 4 794
AK150401.1 - 4 794
AK150670.1 - 4 794
AK151570.1 - 4 794
AK153146.1 - 4 790
AK159145.1 - 4 843
AK159179.1 - 4 794
AK160012.1 - 4 794
AK161610.1 - 4 820
AK164433.1 - 4 794
AK165985.1 - 4 794
AK166637.1 - 4 820
AK178597.1 - 1 317
AK192830.1 - 1 31
AK206902.1 - 1 319
AK212349.1 - 1 319
BC075653.1 - 4 941 30539330
BC088731.1 - 1 575 5340043

View Full Product Details