Gene Transcripts

Gene Symbol
Akap8
Entrez Gene ID
56399
Gene Name
A kinase (PRKA) anchor protein 8
Gene Aliases
1200016A02Rik, AA673585, AKAP-8, AKAP95, AU015639
Chromosome Location
Chr. 17: 32303672 - 32321327 on Build GRCm38
UniGene ID
Mm.328945
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB028920.1 - 14 2025
AK004801.1 - 14 2031
AK077864.1 - 1 15
AK089092.1 - 14 2038
BC042602.1 - 14 2908
BC054433.1 - 12 2605
BC060170.1 - 14 2905
BC060209.1 - 13 2521
BC078448.1 - 13 2411
BC125521.1 - 14 2021 40130036
BC137746.1 - 14 2021 8860765

View Full Product Details