Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190161.1 | NP_001177090.1 | 4 | 216 | |
NM_019976.3 | NP_064360.1 | 4 | 213 | ||
GenBank mRNA | AF156598.1 | - | 4 | 208 | |
AK011197.1 | - | 4 | 188 | ||
AK017302.1 | - | 4 | 155 | ||
AK041835.1 | - | 4 | 163 | ||
AK167415.1 | - | 4 | 159 | ||
AK178652.1 | - | 4 | 208 | ||
BC008994.1 | - | 1 | 132 | 3156760 | |
BC019165.1 | - | 4 | 171 | 3963608 | |
BY080522.1 | - | 4 | 213 | ||
BY231608.1 | - | 4 | 216 |