Gene Transcripts

Gene Symbol
C1qtnf1
Entrez Gene ID
56745
Gene Name
C1q and tumor necrosis factor related protein 1
Gene Aliases
1600017K21Rik, CTRP1, Zsig37
Chromosome Location
Chr. 11: 118428457 - 118451782 on Build GRCm38
UniGene ID
Mm.23845
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204129.1 NP_001191058.1 3 332
NM_001204130.1 NP_001191059.1 3 244
NM_019959.3 NP_064343.1 3 322
GenBank mRNA AF192499.1 - 3 270
AK005484.1 - 3 291
AK145104.1 - 3 245
BC037636.1 - 2 237
BC049894.1 - 3 273 4018549
CJ056741.1 - 3 322
DQ002394.1 - 2 216

View Full Product Details